Synthware™ Buchner Filter Funnels
Used to filter particulates from a variety of substances
Supplier:  Synthware™ F603030X
 View more versions of this product
Catalog No. 31501520
Description
Description
  • Sealed in glass fritted disc of listed size and porosity
  • Porosity: XC-Extra Coarse, C-Coarse, M-Medium, F-Fine
  • Not recommended for filtration where pressure would exceed 15 psi
Specifications
Specifications
Buchner filter funnel
30 mL
30mm
Extra Coarse