Synthware™ Addition Funnels with Pressure Equalizing Arm and PTFE Stopcock
Cylindrical style addition funnel with pressure equalizing arm and PTFE Stopcock
Supplier: Synthware™ F674500
Description
- Top outer and lower inner standard taper joints of same size
Specifications
Pressure Equalizing Funnel with PTFE stopcock | |
500 mL | |
Stopcock bore: 4mm | |
24/40 |