Synthware™ Addition Funnels with Pressure Equalizing Arm and 1:5 PTFE Stopcock
Cylindrical style addition funnel with pressure equalizing arm and PTFE stopcock
Supplier: Synthware™ F624125
Description
- Top outer and lower inner standard taper joints of same size
Specifications
Pressure equalizing funnel | |
125 mL | |
24/40 |