Synthware™ Addition Funnels with High Vacuum Valve
Used for filling containers
Supplier:  Synthware™ F551060
 View more versions of this product
Catalog No. 31501566
Description
Description
  • Upper outer and lower inner standard taper joints of same size
  • Supplied with two 0-4 mm high vacuum valves with PTFE O-ring at tip and PTFE wiper to protect Viton O-ring on shaft
Specifications
Specifications
Addition funnel
100 mL
Glass
14/20