Please login to your online account to display your discounted pricing

Synthetic Fresh Water, Very Soft, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Synthetic Fresh Water
Name Note Very Soft
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 144-55-8
CAS Min % 0.0011
CAS Max % 0.0011
CAS 10101-41-4
CAS Min % 7.0E-4
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

8661-1

 
ricca chemical company
RI86611
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

8661-32

 
ricca chemical company
RI866132
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 4L
Chemical Name or Material Synthetic Fresh Water
CAS Max % 7.0E-4
CAS 7487-88-9
CAS Min % 7.0E-4
Packaging Poly natural
CAS 7447-40-7
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 144-55-8
CAS Max % 0.0011
CAS 10101-41-4
CAS Min % 7.0E-4
CAS Max % 7.0E-4
Name Note Very Soft
CAS Min % 0.0011