Please login to your online account to display your discounted pricing

Synthetic Fresh Water, Very Hard, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Synthetic Fresh Water
Name Note Very Hard
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 144-55-8
CAS Min % 0.04
CAS Max % 0.04
CAS 10101-41-4
CAS Min % 0.02
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

8665-1

 
ricca chemical company
RI86651
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

8665-32

 
ricca chemical company
RI866532
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 4L
Chemical Name or Material Synthetic Fresh Water
CAS Max % 0.02
CAS 7487-88-9
CAS Max % 0.02
Packaging Poly natural
CAS 7447-40-7
CAS Min % 0.0015
CAS Max % 0.0015
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS Min % 0.04
CAS 10101-41-4
CAS Min % 0.02
CAS Min % 0.02
Name Note Very Hard
CAS 144-55-8
CAS Max % 0.04