Please login to your online account to display your discounted pricing

Synthetic Fresh Water, Soft, Ricca Chemical

$89.61 - $89.61

Chemical Identifiers

Chemical Name or Material Synthetic Fresh Water
Name Note Soft
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 144-55-8
CAS Min % 0.0047
CAS Max % 0.0049
CAS 10101-41-4
CAS Min % 0.0028
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

8662-32

 
ricca chemical company
RI866232
Poly natural 1L Each for $89.61

8662-1

 
ricca chemical company
RI86621
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 1L
CAS 7732-18-5
CAS 7487-88-9
CAS Max % 0.0030
CAS 7447-40-7
CAS Min % 0.0028
CAS Max % 0.0030
CAS Min % 98.0
CAS Max % 100.0
CAS Max % 0.0049
CAS Min % 0.0047
CAS Min % 1.0E-4
CAS Max % 1.0E-4
Chemical Name or Material Synthetic Fresh Water
CAS Min % 0.0028
Packaging Poly natural
CAS 10101-41-4
CAS 144-55-8
Name Note Soft