missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Synthetic Fresh Water, Moderately Hard, Ricca Chemical

Chemical Identifiers

CAS 7732-18-5,144-55-8,10101-41-4,7487-88-9,7447-40-7
CAS Min % 98.0,0.01,0.01,0.01,4.0E-4
CAS Max % 100.0,0.01,0.01,0.01,4.0E-4
Physical Form Liquid
Chemical Name or Material Synthetic Fresh Water, Moderately Hard
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
8663-1 SDS Ricca Chemical Company
RI86631
Poly natural 4L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
8663-32 Ricca Chemical Company
RI866332
Poly natural 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,144-55-8,10101-41-4,7487-88-9,7447-40-7
100.0,0.01,0.01,0.01,4.0E-4
Synthetic Fresh Water, Moderately Hard
98.0,0.01,0.01,0.01,4.0E-4
Liquid

Specifications

Liquid
4L
100.0,0.01,0.01,0.01,4.0E-4
Poly natural
Clear colorless
7732-18-5,144-55-8,7447-40-7,10101-41-4,7487-88-9
0.01,0.01,4.0E-4,98.0,0.01
Synthetic Fresh Water, Moderately Hard