Please login to your online account to display your discounted pricing

Synthalin sulfate, Tocris


Non-competitive NMDA antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS063510

 View more versions of this product

Catalog No. 063510

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 354.47
CAS 182285-12-7
Assay Percent Range >98%
Chemical Name or Material Synthalin sulfate
Molecular Formula C12H28N6·H2SO4
Synonym N, N'-1, 10-Decanediylbisguanidine sulfate
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material Synthalin sulfate
Assay Percent Range >98%
Molecular Formula C12H28N6·H2SO4
Formula Weight 354.47
Synonym N, N'-1, 10-Decanediylbisguanidine sulfate
Accessories

Store at Room Temperature