Please login to your online account to display your discounted pricing

Synthalin sulfate, Tocris

Non-competitive NMDA antagonist

$146.74 - $618.87

Chemical Identifiers

Chemical Name or Material Synthalin sulfate
Assay Percent Range >98%
Molecular Formula C12H28N6·H2SO4
Formula Weight 354.47
Synonym N, N'-1, 10-Decanediylbisguanidine sulfate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

063510

 
r & d systems
RDS063510
10mg Each for $146.74

063550

 
r & d systems
RDS063550
50mg Each for $618.87
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 354.47
CAS 182285-12-7
Assay Percent Range >98%
Chemical Name or Material Synthalin sulfate
Molecular Formula C12H28N6·H2SO4
Synonym N, N'-1, 10-Decanediylbisguanidine sulfate
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature