Please login to your online account to display your discounted pricing

Synaptophysin Mouse anti-Human, eFluor 615, Clone: EP10, eBioscience™

Mouse Monoclonal Antibody

$175.45 - $413.25

Specifications

Antigen Synaptophysin
Clone EP10
Host Species Mouse
Species Reactivity Armenian Hamster, Golden Syrian Hamster, Human
Applications Immunocytochemistry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50245825

 
life technologies
LS42652580
25μg Each for $175.45

50245826

 
life technologies
LS42652582
100μg Each for $413.25
Description & Specifications

Specifications

Antigen Synaptophysin
Clone EP10
Host Species Mouse
Species Reactivity Armenian Hamster, Golden Syrian Hamster, Human
Applications Immunocytochemistry
Applications Immunohistochemistry
Regulatory Status RUO
Conjugate eFluor 615
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

This EP10 monoclonal antibody reacts with human Synaptophysin, which is also known as Major Synaptic Vesicle Protein p38. This 38kDa integral membrane protein is expressed on neuronal presynaptic vesicles in the brain, spinal cord, retina, neuromuscular junctions, and adrenal medulla. Synaptophysin is also expressed in neuroendocrine tumors and neuroblastomas. Synaptophysin has been reported to play a role in synaptic vesicle trafficking.

The EP10 antibody recognizes human and hamster synaptophysin but does not recognize mouse and rat synaptophysin.