missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Synaptophysin Mouse anti-Human, Clone: EP10, eBioscience™

Mouse Monoclonal Antibody

$104.28 - $328.68

Specifications

Antigen Synaptophysin
Clone EP10
Host Species Murine
Gene Alias Major Synaptic Vesicle Protein p38
Applications Immunocytochemistry,Immunohistochemistry (Formalin/Paraffin),Immunoprecipitation,Western Blot
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013487 Life Technologies
LS14652580
25μg Each for $104.28
5013488 Life Technologies
LS14652582
100μg Each for $328.68
Specifications

Description

This EP10 monoclonal antibody reacts with human Synaptophysin, which is also known as Major Synaptic Vesicle Protein p38. This 38kDa integral membrane protein is expressed on neuronal presynaptic vesicles in the brain, spinal cord, retina, neuromuscular junctions, and adrenal medulla. Synaptophysin is also expressed in neuroendocrine tumors and neuroblastomas. Synaptophysin has been reported to play a role in synaptic vesicle trafficking.

The EP10 antibody recognizes human and hamster synaptophysin but does not recognize mouse and rat synaptophysin.
Description & Specifications

Specifications

Synaptophysin
Murine
Immunocytochemistry,Immunohistochemistry (Formalin/Paraffin),Immunoprecipitation,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
EP10
Major Synaptic Vesicle Protein p38
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer