missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Synaptophysin Ab-4, Rabbit Polyclonal Antibody

Provide accurate, reproducible results Thermo Scientific™ Synaptophysin Ab-4, Rabbit Polyclonal Antibody.

$356.96 - $1,406.41

Specifications

Antigen Synaptophysin Ab-4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 6
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
RB1461P View Documents Thermo Scientific™
RB1461P
Purified Ab with BSA and Azide 1mL Each for $1,229.49
 
RB1461P0 View Documents Thermo Scientific™
RB1461P0
Purified Ab with BSA and Azide 100μL Each for $356.96
 
RB1461P1 View Documents Thermo Scientific™
RB1461P1
Purified Ab with BSA and Azide 500μL Each for $1,071.51
 
RB1461P1ABX View Documents Thermo Scientific™
RB1461P1ABX
Ab without BSA/Azide (Frozen) 500μL Each for $1,083.90
 
RB1461PABX View Documents Thermo Scientific™
RB1461PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,406.41
 
RB1461R7 View Documents Thermo Scientific™
RB1461R7
Ready-to-use for IHC (UVLP) 7mL Each for $654.80
 
Description

Description

Provide accurate, reproducible results Thermo Scientific™ Synaptophysin Ab-4, Rabbit Polyclonal Antibody.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Rat. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide from C-terminus of human synaptophysin

Molecular Weight: 38kDa

Positive Control: IMR-5 cells, Pancreas or Pheochromocytoma or GI tumor

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Synaptophysin Ab-4
Polyclonal
Rabbit
Cancer and Tumor Biology
A synthetic peptide from C-terminus of human synaptophysin
Western Blot
Unconjugated
RUO
Primary
IgG