Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Synaptophysin Ab-4, Rabbit Polyclonal Antibody


Provide accurate, reproducible results Thermo Scientific™ Synaptophysin Ab-4, Rabbit Polyclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RB1461PABX

 View more versions of this product

Catalog No. RB1461PABX

  • / Each
 

Description & Specifications

Specifications

Host Species Rabbit
Applications Western Blotting
Isotype IgG
Antigen Synaptophysin Ab-4
Format Concentrated
Cross Reactivity Human
Quantity 1mL
Immunogen A synthetic peptide from C-terminus of human synaptophysin
Regulatory Status RUO
Concentration 1mg/mL
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Cancer and Tumor Biology
Conjugate Unlabeled
Type Primary
Classification Polyclonal

Provide accurate, reproducible results Thermo Scientific™ Synaptophysin Ab-4, Rabbit Polyclonal Antibody.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Rat. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide from C-terminus of human synaptophysin

Molecular Weight: 38kDa

Positive Control: IMR-5 cells, Pancreas or Pheochromocytoma or GI tumor

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO