Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Synaptophysin Ab-4, Rabbit Polyclonal Antibody


Provide accurate, reproducible results Thermo Scientific™ Synaptophysin Ab-4, Rabbit Polyclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RB1461P1ABX

 View more versions of this product

Catalog No. RB1461P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen Synaptophysin Ab-4
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Host Species Rabbit
Immunogen A synthetic peptide from C-terminus of human synaptophysin
Isotype IgG
Quantity 500μL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Type Primary
Monoclonal or Polyclonal Polyclonal
Target Species Human

Provide accurate, reproducible results Thermo Scientific™ Synaptophysin Ab-4, Rabbit Polyclonal Antibody.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Rat. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide from C-terminus of human synaptophysin

Molecular Weight: 38kDa

Positive Control: IMR-5 cells, Pancreas or Pheochromocytoma or GI tumor

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO