missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

syn-2-Nitrobenzaldoxime [Deprotecting Agent], TCI America™

$41.25 - $123.75

Chemical Identifiers

CAS 4836-00-4
Molecular Formula C7H6N2O3
Molecular Weight (g/mol) 166.136
MDL Number MFCD00007187
InChI Key IHMGDCCTWRRUDX-VMPITWQZSA-N
PubChem CID 7332808
IUPAC Name (NE)-N-[(2-nitrophenyl)methylidene]hydroxylamine
SMILES C1=CC=C(C(=C1)C=NO)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N04865G View Documents TCI America
N04865G
5g Each for $41.25
 
N048625G View Documents TCI America
N048625G
25g Each for $123.75
 
Specifications

Chemical Identifiers

4836-00-4
166.136
IHMGDCCTWRRUDX-VMPITWQZSA-N
(NE)-N-[(2-nitrophenyl)methylidene]hydroxylamine
C7H6N2O3
MFCD00007187
7332808
C1=CC=C(C(=C1)C=NO)[N+](=O)[O-]

Specifications

Yellow
5g
C7H6N2O3
IHMGDCCTWRRUDX-VMPITWQZSA-N
(NE)-N-[(2-nitrophenyl)methylidene]hydroxylamine
7332808
Crystal-Powder at 20°C
syn-2-Nitrobenzaldoxime [Deprotecting Agent]
95°C
4836-00-4
MFCD00007187
C1=CC=C(C(=C1)C=NO)[N+](=O)[O-]
166.136
166.14
≥85.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No