missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

sym-Diphenylcarbazone, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$224.36 - $648.14

Chemical Identifiers

Molecular Formula C6H5NHNHCON:NC6H5·C6H5NHNHCONHNHC6H5
Formula Weight 482.55
Chemical Name or Material sym-Diphenylcarbazone
Grade Baker Analyzed™ ACS Reagent
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
02004444 Avantor Performance
K61702
Wide Mouth Amber Glass Bottle 10g Each for $224.36
02004445 Avantor Performance
K61703
Wide Mouth Amber Glass Bottle 25g Each for $648.14
Specifications Description & Specifications

Chemical Identifiers

C6H5NHNHCON:NC6H5·C6H5NHNHCONHNHC6H5
482.55
sym-Diphenylcarbazone
Baker Analyzed™ ACS Reagent

Specifications

Wide Mouth Amber Glass Bottle
482.55
sym-Diphenylcarbazone
C6H5NHNHCON:NC6H5·C6H5NHNHCONHNHC6H5
10g
Baker Analyzed™ ACS Reagent