missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

sym-Diphenylcarbazide, 98%, ACS reagent, ACROS Organics™

Manufacturer:  Acros Organics AC158845000

 View more versions of this product

Catalog No. AC158845000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Indicator
Specifications

Specifications

sym-Diphenylcarbazide
1% max.
(to chromate) passes test
Beige to Pink
500g
98%
1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl
KSPIHGBHKVISFI-UHFFFAOYSA-N
1,3-dianilinourea
8789
242.28
98%
Authentic
Glass bottle
0.05 % max.
173.0°C to 176.0°C
140-22-7
C13H14N4O
Solubility in water: slightly soluble. Other solubilities: soluble in glacial acetic acid, soluble in alcohol and acetone
C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2
242.282
CHEBI:4641
Powder or Flakes
ACS Reagent

Chemical Identifiers

140-22-7
242.282
1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl
CHEBI:4641
C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2
C13H14N4O
KSPIHGBHKVISFI-UHFFFAOYSA-N
8789
1,3-dianilinourea
SDS Documents