Please login to your online account to display your discounted pricing

sym-Diformylhydrazine 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material sym-Diformylhydrazine
Name Note 97%
CAS 628-36-4
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C2H4N2O2
Linear Formula HCONHNHCHO
Formula Weight 88.06
MDL Number MFCD00003275
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC114640250

  SDS
thermo fisher scientific
114640250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC114641000

  SDS
thermo fisher scientific
114641000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 88.06
Physical Form Fine Crystalline Powder
Quantity 25g
Color White to almost white
TSCA TSCA
EINECS Number 211-040-5
RTECS Number LQ8625000
Melting Point 155° to 160°C
Beilstein 02, 93
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 97%
Linear Formula HCONHNHCHO
Assay Percent Range 97%
Chemical Name or Material sym-Diformylhydrazine
Molecular Formula C2H4N2O2
MDL Number MFCD00003275
CAS 628-36-4