Please login to your online account to display your discounted pricing

SYM 2206, Tocris


Non-competitive AMPA antagonist

Quantity 50mg

Manufacturer: r & d systems  096150

 View more versions of this product

Catalog No. 096150

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 366.42
Assay Percent Range >99%
Molecular Formula C20H22N4O3
Chemical Name or Material SYM 2206
CAS 173952-44-8
Synonym (±)-4-(4-Aminophenyl)-1, 2-dihydro-1-methyl-2-propylcarbamoyl-6, 7-methylenedioxyphthalazine
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material SYM 2206
Assay Percent Range >99%
Molecular Formula C20H22N4O3
Formula Weight 366.42
Synonym (±)-4-(4-Aminophenyl)-1, 2-dihydro-1-methyl-2-propylcarbamoyl-6, 7-methylenedioxyphthalazine
Accessories

Store at Room Temperature