missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SYM 2206, Tocris

Non-competitive AMPA antagonist

$232.08 - $956.46

Chemical Identifiers

Assay Percent Range >99%
Molecular Formula C20H22N4O3
Formula Weight 366.42
Synonym (±)-4-(4-Aminophenyl)-1, 2-dihydro-1-methyl-2-propylcarbamoyl-6, 7-methylenedioxyphthalazine
Chemical Name or Material SYM 2206
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
096110 R & D Systems
096110
10mg Each for $232.08
096150 R & D Systems
096150
50mg Each for $956.46
Specifications Description & Specifications

Chemical Identifiers

>99%
366.42
SYM 2206
C20H22N4O3
(±)-4-(4-Aminophenyl)-1, 2-dihydro-1-methyl-2-propylcarbamoyl-6, 7-methylenedioxyphthalazine

Specifications

173952-44-8
10mg
366.42
SYM 2206
>99%
C20H22N4O3
(±)-4-(4-Aminophenyl)-1, 2-dihydro-1-methyl-2-propylcarbamoyl-6, 7-methylenedioxyphthalazine

Safety and Handling

Recommended Storage : Store at Room Temperature