Please login to your online account to display your discounted pricing

SYM 2081, Tocris


Highly selective kainate agonist

Quantity 10mg

Manufacturer: r & d systems  RDS090310

 View more versions of this product

Catalog No. 090310

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 161.16
Assay Percent Range >97%
Molecular Formula C6H11NO4
Recommended Storage Store at Room Temperature
Synonym (2S, 4R)-4-Methylglutamic acid
Chemical Name or Material SYM 2081
CAS 31137-74-3

Specifications

Chemical Name or Material SYM 2081
Assay Percent Range >97%
Molecular Formula C6H11NO4
Formula Weight 161.16
Synonym (2S, 4R)-4-Methylglutamic acid
Accessories

Store at Room Temperature