Please login to your online account to display your discounted pricing

SYM 2081, Tocris

Highly selective kainate agonist

$244.81 - $997.23

Chemical Identifiers

Chemical Name or Material SYM 2081
Assay Percent Range >97%
Molecular Formula C6H11NO4
Formula Weight 161.16
Synonym (2S, 4R)-4-Methylglutamic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

090350

 
r & d systems
RDS090350
50mg Each for $997.23

090310

 
r & d systems
RDS090310
10mg Each for $244.81
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 161.16
Chemical Name or Material SYM 2081
Assay Percent Range >97%
Molecular Formula C6H11NO4
CAS 31137-74-3
Synonym (2S, 4R)-4-Methylglutamic acid
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature