Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Syk Signaling Protein Ab-1, Mouse Monoclonal Antibody


Provide accurate, reproducible results Thermo Scientific™ Syk Signaling Protein Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS322P1ABX

 View more versions of this product

Catalog No. MS322P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen Syk Signaling Protein Ab-1
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Clone 4D10.1
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Host Species Murine
Immunogen Synthetic peptide corresponding to aa 313-339 of human Syk protein
Isotype IgG2a kappa
Quantity 100μL
Regulatory Status RUO
Research Discipline Signal Transduction
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human

Provide accurate, reproducible results Thermo Scientific™ Syk Signaling Protein Ab-1, Mouse Monoclonal Antibody.

Syk, a protein tyrosine kinase (PTK) is expressed in immature T cells, all B cells, basophils, hepatocytes, platelets, and erythrocytes. It functions in some T cell receptor signalling processes in addition to B cell receptor and integrin-mediated signal transduction processes. syk-/syk- mice exhibit defective phagocytosis by macrophages.

Host Species: Mouse

Clone: 4D10.1

Isotype: IgG2a/κ

Species Reactivity: Human. Does not react with pig or mouse. Others not known.

Epitope: aa 313-339

Immunogen: Synthetic peptide corresponding to aa 313-339 of human Syk protein

Molecular Weight: 72kDa

Positive Control: Raji, K562, HL-60, U937 cells or lymphoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native and denatured)
  • in vitro Kinase Assay (Not verified)

Note:

USA: RUO; Int'l: RUO