missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Syk Mouse anti-Human, APC, Clone: 4D10.1, eBioscience™

Mouse Monoclonal Antibody

$161.04 - $389.40

Specifications

Antigen Syk
Clone 4D10.1
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5015287 Life Technologies
LS17669641
25 Tests Each for $161.04
5015288 Life Technologies
LS17669642
100 Tests Each for $389.40
Specifications

Description

This 4D10.1 monoclonal antibody recognizes human Syk, a 72kDa member of the Syk/ZAP-70 family of non-receptor protein tyrosine kinases. Syk is expressed most highly in B cells, with lower expression in immature thymocytes, mast cells, and platelets. Syk can also be detected in fibroblasts, epithelial cells, hepatocytes, endothelial cells, and neuronal cells. This protein is a major component of signaling cascades downstream of the B and T cell antigen receptors and plays an essential role in lymphocyte development. Upon recruitment to immunoreceptor tyrosine-based activation motifs (ITAMs) on the antigen receptors, Syk is activated by phosphorylation on multiple tyrosines. Once activated, Syk phosphorylates proteins such as phospholipase C gamma and BLNK/SLP-65. Finally, abnormal Syk expression has been linked to tumor cell migration and invasion in several cancers.
Description & Specifications

Specifications

Syk
Murine
Flow Cytometry (Intracellular Staining)
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test
4D10.1
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer