Please login to your online account to display your discounted pricing

Tocris™ SX 011

p38 MAPK inhibitor

$252.39 - $1,050.42

Specifications

Product Type SX 011
Inhibitors SX 011
Molecular Weight 483.96
Purity >98
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

363910R

 
tocris™
RDS363910
10mg Each for $252.39

363950R

 
tocris™
RDS363950
50mg Each for $1,050.42
Description & Specifications

Specifications

Product Type SX 011
Inhibitors SX 011
Molecular Weight 483.96
Purity >98

Potent p38α inhibitor (IC50 = 9nM). Also inhibits p38β and JNK-2 (IC50 values are 90nM and 100nM respectively). Displays no significant activity at p38γ, p38δ, ERK-2 and JNK-1.

  • Chemical Name: 6-Chloro-5-[[4-[(4-fluorophenyl)methyl]-1-piperidinyl]carbonyl-N,N,1-trimethyl-α-oxo-1H-indole-3-acetamide
  • Formula: C26H27ClFN3O3
  • CAS Number: 309913-42-6
  • Solubility: Soluble to 100mM in DMSO and to 100mM in ethanol
  • Purity: >98