Swing-Away Eyewash
Save valuable countertop space.
Manufacturer:  Encon Safety Products 01040128
Catalog No. 29-635-2227
Description
Description
  • Water flows continuously with a single motion
  • Pull arm over the sink to turn on, and return it to shut off
  • Install on countertops up to 2 in. thick
  • Can be mounted on either side of sink
  • Acetal heads are resistant to acetone
  • 1/2 in. NPT supply connection
Compliance
Meets ANSI Z358.1-2009 standards
Specifications
Specifications
Swing away eyewash
ANSI Z358.1-2009
Acetal spray head, 10.5 in. arm, NPT supply connection, actuation graphic
Right Hand Countertop