missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Swift™ Tri-Power Stereomicroscopes

Like no other microscopes on the market, this unique series outperforms standard stereomicroscopes.

$550.25 - $550.25

Specifications

Axis Type Rack and pinion with slip clutch
Certifications/Compliance UC/cUL
Quantity 1
Eyepiece W10X
Height (English) 11.5 in.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Magnification Power Objective Types Price Quantity  
Catalog Number Mfr. No. Magnification Power Objective Types Price Quantity  
S19618 Fisher Scientific
M27B124EMD
1x, 2x and 4x 1x, 2x and 4x Each for $550.25
Specifications

Description

  • Three magnification settings on each scope
  • Wide-field, paired eyepieces; W10X, locked-on
  • Adjustable interpupillary distance and diopter correction
  • Built-in illumination, both top and bottom LED illumination
  • Two stage plates: one reversible, opaque black and white; and one frosted transparent specimen (100mm dia.)
  • Working distance, 50 or 30mm

Compliance

UC/CUL approved

Warranty

Limited lifetime on all defective parts and workmanship

Description & Specifications

Specifications

Rack and pinion with slip clutch
1
11.5 in.
Reversible black and white and frosted stage plates, Vinyl dust cover, eyeshields and instruction manual
Lifetime
13.4kg
UC/cUL
W10X
29.21cm
LED
6.1 lb.