Please login to your online account to display your discounted pricing
Swift™ M3700 WiFi Series Advanced Microscope
Built-in high-resolution streaming WiFi camera sets this microscope apart from others.
Manufacturer: fisher scientific  M3702C3DGLXEMD
 View more versions of this product
Catalog No. S04749
  • / Each
Description & Specifications
Specifications
Magnification Power 40xD, 100xD, 400xRD semi plan
Voltage 110/220V
Axis Type Coaxial
Objective Types 4xD, 10xD, 40xRD semi plan
Camera Type 1.3MP 1280 x 1024 pixel CMOS
Condenser Type 1.25 Abbe
Head Style Monocular
Includes MotiConnect app, calibration slides, macro tube, 12mm focusing lens
Design Type Mechanical
Diaphragm Iris
Eyepiece W10XD, 18mm
Lighting Type Corded LED with dimmer switch
Port Type USB
Warranty Lifetime
Software Type Motic