missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Swift™ M10D Series Digital Microscopes with High-Resolution Camera

Built-in, high-resolution 3 MP camera displays live images and captures pictures at 2048 × 1536 resolution.

Manufacturer:  Swift™ M10DBMPEMD

 View more versions of this product

Catalog No. S95264L



Includes: Motic Images Plus imaging software. Software is Windows™ XP™, Vista™ compatible and Mac™ OSX™ compatible.

Description

Description

  • Binocular head available with semi-plan, plan or multi-phase objectives (4XD, 10XD, 40XRD, 100XRD)
  • Internal camera allows use of eyepiece at the same time as camera
  • Illuminated by a corded LED system with dimmer switch
  • USB output provides a direct connection to computer
  • Built-in low-drive mechanical stage
  • Abbe 1.25 DIN rack and pinion condenser, iris diaphragm
  • 20mm eyepiece

Warranty

Limited lifetime warranty on microscope and one year limited on camera.

Specifications

Specifications

Binocular
3w LED with dimmer switch
36.9cm
Coaxial coarse and fine
1.25 DIN Abbe rack and pinion
Iris
Motic
14kg
Lifetime
Motic™ images plus software
14.25 in.
110/220V
Mechanical
4xD Plan, 10xD Ph, 40xRD Ph, 100xRD Ph
4xD plan, 10xD Ph, 40xRD Ph, 100xRD Ph
USB
30.9 lb.
SDS
Documents