missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ SV40 Large T Antigen Ab-2, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ SV40 Large T Antigen Ab-2, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ Lab Vision™ MS1833P1ABX

 View more versions of this product

Catalog No. MS1833P1ABX


 

Specifications

Description

Provide accurate, reproducible results with the Thermo Scientific™ SV40 Large T Antigen Ab-2, Mouse Monoclonal Antibody.

The simian virus 40 large T antigen plays important role in SV40 transfection and transformation of the cells. It binds to Rb and p53 suppressor proteins leading to the inactivation of suppressor pathway.

Host Species: Mouse

Clone: PAb108

Isotype: IgG2a1

Species Reactivity: SV40 virus.

Epitope: N-terminal

Immunogen: SV40-transformed mouse line B4

Molecular Weight: 94kDa

Positive Control: SV40-transfected cells

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

SV40 Large T Antigen Ab-2
Monoclonal
1mg/mL
Concentrated
SV40 transformed mouse line B4
RUO
Primary
SV40 Virus
Immunofluorescence,Immunoprecipitation,Western Blotting
PAb108
Unlabeled
Murine
100μL
Infectious Disease
Monoclonal