Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ SV40 Large T Antigen Ab-2, Mouse Monoclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ SV40 Large T Antigen Ab-2, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1833PABX

 View more versions of this product

Catalog No. MS1833PABX

  • / Each
 

Description & Specifications

Specifications

Antigen SV40 Large T Antigen Ab-2
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Clone PAb108
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity SV40 Virus
Format Concentrated
Host Species Murine
Immunogen SV40 transformed mouse line B4
Quantity 200μL
Regulatory Status RUO
Research Discipline Infectious Disease
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species SV40 Virus

Provide accurate, reproducible results with the Thermo Scientific™ SV40 Large T Antigen Ab-2, Mouse Monoclonal Antibody.

The simian virus 40 large T antigen plays important role in SV40 transfection and transformation of the cells. It binds to Rb and p53 suppressor proteins leading to the inactivation of suppressor pathway.

Host Species: Mouse

Clone: PAb108

Isotype: IgG2a1

Species Reactivity: SV40 virus.

Epitope: N-terminal

Immunogen: SV40-transformed mouse line B4

Molecular Weight: 94kDa

Positive Control: SV40-transfected cells

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO