Please login to your online account to display your discounted pricing

Survivin (BIRC5) Mouse anti-Human, Clone: STLALYV, eBioscience™

Mouse Monoclonal Antibody

$191.40 - $427.75

Specifications

Antigen Survivin (BIRC5)
Clone STLALYV
Host Species Mouse
Gene Alias Birc5
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122774

 
life technologies
LS14917680
25μg Each for $191.40

501122775

 
life technologies
LS14917682
100μg Each for $427.75
Description & Specifications

Specifications

Antigen Survivin (BIRC5)
Clone STLALYV
Host Species Mouse
Gene Alias Birc5
Species Reactivity Human
Applications Immunocytochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Microscopy
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

This STLALYV monoclonal antibody recognizes human survivin (also known as Birc5). Survivin is a member of the inhibitor of apoptosis (IAP) family that also includes XIAP, c-IAP1, c-IAP2, and NAIP. Survivin functions to inhibit caspase activation, thereby leading to negative regulation of apoptosis. Survivin also plays a critical role in mitosis and cytokinesis where it is expressed only during G2/M and localizes to the mitotic spindle and is found at the cleavage furrow/midplate. Thus, survivin is highly expressed in tumors and in fetal tissue, but is absent in terminally-differentiated cells and non-proliferating cells. Deletion of survivin in developing thymocytes as well as in mature T cells showed that survivin is critical for cell division in lymphoid cells but not for inhibition of survival; the same is true for B lymphocytes.