Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Survivin Ab-6, Rabbit Polyclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Survivin Ab-6, Rabbit Polyclonal Antibody.

$274.45 - $1,353.14

Specifications

Antigen Survivin Ab-6
Concentration 1mg/mL
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

RB1629P

 
thermo scientific™ lab vision™
RB1629P
Purified Ab with BSA and Azide 1mL Each for $1,270.22

RB1629P0

 
thermo scientific™ lab vision™
RB1629P0
Purified Ab with BSA and Azide 100µL Each for $274.45

RB1629P1

 
thermo scientific™ lab vision™
RB1629P1
Purified Ab with BSA and Azide 500µL Each for $752.41

RB1629P1ABX

 
thermo scientific™ lab vision™
RB1629P1ABX
Ab without BSA/Azide (Frozen) 500µL Each for $844.49

RB1629PABX

 
thermo scientific™ lab vision™
RB1629PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,353.14
Description & Specifications

Specifications

Antigen Survivin Ab-6
Concentration 1mg/mL
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Rabbit
Regulatory Status RUO
Research Discipline Apoptosis
Type Primary
Immunogen A synthetic peptide derived from the C-terminus of survivin
Isotype IgG

Provide accurate, reproducible results with the Thermo Scientific™ Survivin Ab-6, Rabbit Polyclonal Antibody.

Survivin, a new inhibitor of apoptosis (IAP) protein, is expressed in the G2/M phase of the cell cycle in a cycle-regulated manner. At the beginning of mitosis, survivin associates with microtubules of the mitotic spindle in a specific and saturable reaction that is regulated by microtubule dynamics. Disruption of survivin-microtubule interactions results in loss of survivin's anti-apoptosis function and increased caspase-3 activity, a mechanism involved in cell death, during mitosis. It indicates that survivin may counteract a default induction of apoptosis in G2/M phase. The overexpression of survivin in cancer may overcome this apoptotic checkpoint and favour aberrant progression of transformed cells through mitosis.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Mouse. Others not tested.

Epitope: C-terminal

Immunogen: A synthetic peptide derived from the C-terminus of survivin

Molecular Weight: 16.5kDa

Positive Control: Stomach

Cellular Localization: Cytoplasmic and Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO