Please login to your online account to display your discounted pricing

Thermo Scientific™ Survivin Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Survivin Ab-1, Mouse Monoclonal Antibody.

$242.00 - $1,278.80

Specifications

Antigen Survivin Ab-1
Clone 8E2
Applications Immunofluorescence
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS1201P

 
thermo scientific™
MS1201P
200μg/mL 1mL Each for $1,192.92

MS1201P0

 
thermo scientific™
MS1201P0
200μg/mL 100µL Each for $242.00

MS1201P1

 
thermo scientific™
MS1201P1
200μg/mL 500µL Each for $773.22

MS1201P1ABX

 
thermo scientific™
MS1201P1ABX
1mg/mL 100µL Each for $845.90

MS1201PABX

 
thermo scientific™
MS1201PABX
1mg/mL 200µL Each for $1,278.80
Description & Specifications

Specifications

Antigen Survivin Ab-1
Clone 8E2
Applications Immunofluorescence
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Rat
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Apoptosis
Type Primary
Immunogen Full length recombinant human survivin protein
Isotype IgG2a kappa

Provide accurate, reproducible results with the Thermo Scientific™ Survivin Ab-1, Mouse Monoclonal Antibody.

Research Areas: Apoptosis; Survivin, a new inhibitor of apoptosis (IAP) protein, is expressed in the G2/M phase of the cell cycle in a cycle-regulated manner. At the beginning of mitosis, survivin associates with microtubules of the mitotic spindle in a specific and saturable reaction that is regulated by microtubule dynamics. Disruption of survivin-microtubule interactions results in loss of survivin's anti-apoptosis function and increased caspase-3 activity, a mechanism involved in cell death, during mitosis. It indicates that survivin may counteract a default induction of apoptosis in G2/M phase. The overexpression of survivin in cancer may overcome this apoptotic checkpoint and favour aberrant progression of transformed cells through mitosis.

Host Species: Mouse

Clone: 8E2

Isotype: IgG1/κ

Species Reactivity: Human and Rat. Others not known.

Epitope: aa 1-99

Immunogen: Full-length recombinant human survivin protein

Molecular Weight: 16.5kDa

Positive Control: HeLa cells

Cellular Localization: Cytoplasmic and nuclear

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting (Not suitable)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO