Please login to your online account to display your discounted pricing

Surrogate Standard Solution

$74.60 - $74.60

Chemical Identifiers

Chemical Name or Material Surrogate Standard Solution
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

USPPS3901

 
ultra scientific
801175029595
Ampule 1x1mL Each for $74.60
Description & Specifications

Specifications

Quantity 1x1mL
Concentration or Composition (by Analyte or Components) 2, 3-Dibromopropionic Acid at 1000μg/mL in Methyl tert-Butyl Ether.
Methods For EPA Methods 552.1, 552.2
Chemical Name or Material Surrogate Standard Solution
Packaging Ampule