Please login to your online account to display your discounted pricing

Surrogate Standard Mixture (For EPA 8260)

$113.77 - $113.77

Chemical Identifiers

Chemical Name or Material Surrogate Standard Mixture
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

US-STM-330N-1

 
ultra scientific
STM330N1
Ampule 1 × 1mL Each for $113.77
Description & Specifications

Specifications

Quantity 1 × 1mL
Concentration or Composition (by Analyte or Components) Each component at 2000μg/mL in Methanol:
4-Bromofluorobenzene
Dibromofluoromethane
Toluene-d8
Methods For EPA Method 8260
Chemical Name or Material Surrogate Standard Mixture
Packaging Ampule