Please login to your online account to display your discounted pricing
Surgical Design Scalpel Handles and Blades
Standard surgeon's scalpel handles are stainless steel
$11.81 - $15.81
Specifications
Design Surgical
Material Stainless Steel
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    
22-079657
  Accessories
surgical design
27057
3 Each for $15.81
22-079681
surgical design
27081
10 Pack of 12 Each for $11.81
22-079665
surgical design
27165
4 Each for $15.81
Description & Specifications
Specifications
Design Surgical
Material Stainless Steel
  • Blades are high quality surgical steel, tempered and hardened for longer and sharper service with the correct balance of rigidity and flexibility
  • Size 3 and 7 handles use blade 10 or 11; size 4 handle uses blade 22