Please login to your online account to display your discounted pricing

Suplatast tosylate, Tocris™

Th2 cytokine synthesis inhibitor; antiasthmatic

$142.08 - $598.33

Chemical Identifiers

Chemical Name or Material Suplatast tosylate
Assay Percent Range >99%
Molecular Formula C16H26NO4S·C7H7O3S
Formula Weight 499.64
Synonym [3-[[4-(3-Ethoxy-2-hydroxypropoxy)phenyl]amino]-3-oxopropyl]dimethylsulfonium 4-methylbenzenesulfonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

354350

 
r & d systems
RDS354350
50mg Each for $598.33

354310

 
r & d systems
RDS354310
10mg Each for $142.08
Description & Specifications

Specifications

Formula Weight 499.64
Quantity 50mg
Chemical Name or Material Suplatast tosylate
Assay Percent Range >99%
Molecular Formula C16H26NO4S·C7H7O3S
CAS 94055-76-2
Synonym [3-[[4-(3-Ethoxy-2-hydroxypropoxy)phenyl]amino]-3-oxopropyl]dimethylsulfonium 4-methylbenzenesulfonate
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature