missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Suplatast Tosylate 98.0+%, TCI America™
Supplier: TCI America S0875100MG
Specifications
Suplatast Tosylate | |
87°C | |
MFCD00867604 | |
suplatast tosilate, suplatast tosylate, suplatast, suplatast tosilate inn, ccris 6596, suplatastum tosilas inn-latin, tosilate de suplatast inn-french, tosilato de suplatast inn-spanish, +--2-4-3-ethoxy-2-hydroxypropoxy phenyl carbamoyl ethyldimethylsulfonium p-tosylate | |
CC1=CC=C(C=C1)S([O-])(=O)=O.CCOCC(O)COC1=CC=C(NC(=O)CC[S+](C)C)C=C1 | |
499.64 | |
499.64 | |
Crystalline Powder |
94055-76-2 | |
C23H33NO7S2 | |
100 mg | |
RYVJQEZJUFRANT-UHFFFAOYNA-N | |
(2-{[4-(3-ethoxy-2-hydroxypropoxy)phenyl]carbamoyl}ethyl)dimethylsulfanium 4-methylbenzene-1-sulfonate | |
71773 | |
≥98.0% (HPLC,N) |
Chemical Identifiers
94055-76-2 | |
499.64 | |
RYVJQEZJUFRANT-UHFFFAOYNA-N | |
71773 | |
CC1=CC=C(C=C1)S([O-])(=O)=O.CCOCC(O)COC1=CC=C(NC(=O)CC[S+](C)C)C=C1 |
C23H33NO7S2 | |
MFCD00867604 | |
suplatast tosilate, suplatast tosylate, suplatast, suplatast tosilate inn, ccris 6596, suplatastum tosilas inn-latin, tosilate de suplatast inn-french, tosilato de suplatast inn-spanish, +--2-4-3-ethoxy-2-hydroxypropoxy phenyl carbamoyl ethyldimethylsulfonium p-tosylate | |
(2-{[4-(3-ethoxy-2-hydroxypropoxy)phenyl]carbamoyl}ethyl)dimethylsulfanium 4-methylbenzene-1-sulfonate |
Safety and Handling
RTECSNumber : WR7906000
TSCA : No