Corning™ Supersome™ Ultra™ HEK Control Cytosol Vesicle
Used for study of drug metabolism. Supersome™ Ultra™ HEK Control Cytosol Vesicle is used in high throughput assays.
Supplier:  Corning™ 456800
Catalog No. CG456800
Description
Description
  • Mammalian cell expression system
  • High throughput models
  • Ideal for use in pre screening assays
  • Measures drug candidates ability
  • Can be used with non radiolabelled isotopes
  • Measurres drug stimulated vanadate-sensitive ATPase activity
  • Mammalian cell expression system
  • Higher uptake activity
  • Lower background
  • Eliminates biosafety concern from baculovirus
  • Improve in vitro to in vivo prediction
Specifications
Specifications
-80°C
Absorption and transport
HEK Cell Cytosolic Vesicle
60 months
Recombinant metabolic enzyme
0.5 mL
0.5 mL
HEK Cell Cytosolic Vesicle