Learn More
Thermo Scientific™ SuperSignal™ West Pico Rabbit IgG Detection Kit
Combines SuperSignal West Pico substrate with HRP-conjugated rabbit or mouse secondary antibodies for a convenient, easy-to-use Western blot detection kit.
Supplier: Thermo Scientific™ LSG34083
Description
Thermo Scientific™ SuperSignal Western Blotting Kits are ideal for busy researchers or people new to Western Blotting.
All the components of these Western blotting kits are optimized for compatibility the SuperSignal West Pico Chemiluminescent Substrate and are available for the detection of mouse and rabbit secondary primary antibodies. All SuperSignal Western Blotting kits come with complete, detailed instructions for those new to the technique of Western Blotting and abbreviated protocols for the experienced user. The kits are also available in a standard format (substrate and optimized HRP-conjugates) or a complete format which also includes the Thermo Scientific SuperBlock Buffer and BupH Tris Buffered Saline.
Highlights:
Standard and complete kits – choose kits with just secondary antibody and chemiluminescent substrate or kits that also include blocking buffer and wash buffer
Mouse and rabbit kits – choose kits with goat anti-mouse IgG or goat anti-rabbit IgG secondary antibody conjugates
Instructions – complete, detailed manual with recommended antibody dilutions
Ordering Information:
Each kit provides sufficient reagents to process 50 miniblots or 4000cm2 of membrane.
Includes:
Standard Detection Kits Include: West Pico Substrate (luminol/enhancer solution and stable peroxide solution), HRP-conjugated anti-rabbit IgG, anti-mouse IgG, or biotin-binding protein. Complete Detection Kits also include SuperBlock Blocking Buffer and TBS wash buffer.
Specifications
SuperSignal West Pico Rabbit IgG Detection Kit | |
Chemiluminescent | |
Nitrocellulose, PVDF | |
HRP | |
Rabbit | |
Sufficient For: ∼62 mini blots probed with rabbit primary antibodies • SuperSignal West Pico Luminol/Enhancer Solution, 250 mL • SuperSignal West Pico Stable Peroxide Solution, 250 mL • Peroxidase Conjugated Goat anti-Rabbit IgG (H + L), 2 mL |
Rabbit IgG Detection Kit | |
Western Blot | |
SuperSignal™ | |
Horseradish Peroxidase | |
50 mini blots (4000 sq. cm of membrane) |
For Research Use Only. Not for use in diagnostic procedures.