Thermo Scientific™ SuperSignal™ Western Blotting Kits
Combines SuperSignal West Pico substrate with HRP-conjugated rabbit or mouse secondary antibodies for a convenient, easy-to-use Western blot detection kit.
$532.92 - $742.96
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI34081 Thermo Scientific™
PI34081
SuperSignal West Pico Complete Mouse IgG Detection Kit Pack for $742.96
PI34082 Thermo Scientific™
PI34082
SuperSignal West Pico Mouse IgG Detection Kit Pack for $532.92
PI34083 Thermo Scientific™
PI34083
SuperSignal West Pico Rabbit IgG Detection Kit Pack for $532.92
PI34084 Thermo Scientific™
PI34084
SuperSignal West Pico Complete Rabbit IgG Detection Kit Pack for $742.96
Specifications
Description
Thermo Scientific™ SuperSignal Western Blotting Kits are ideal for busy researchers or people new to Western Blotting.
All the components of these Western blotting kits are optimized for compatibility the SuperSignal West Pico Chemiluminescent Substrate and are available for the detection of mouse and rabbit secondary primary antibodies. All SuperSignal Western Blotting kits come with complete, detailed instructions for those new to the technique of Western Blotting and abbreviated protocols for the experienced user. The kits are also available in a standard format (substrate and optimized HRP-conjugates) or a complete format which also includes the Thermo Scientific SuperBlock Buffer and BupH Tris Buffered Saline.
Highlights:
  • Standard and complete kits – choose kits with just secondary antibody and chemiluminescent substrate or kits that also include blocking buffer and wash buffer
  • Mouse and rabbit kits – choose kits with goat anti-mouse IgG or goat anti-rabbit IgG secondary antibody conjugates
  • Instructions – complete, detailed manual with recommended antibody dilutions
Ordering Information:
Each kit provides sufficient reagents to process 50 miniblots or 4000cm2 of membrane.
Includes:
Standard Detection Kits Include: West Pico Substrate (luminol/enhancer solution and stable peroxide solution), HRP-conjugated anti-rabbit IgG, anti-mouse IgG, or biotin-binding protein. Complete Detection Kits also include SuperBlock Blocking Buffer and TBS wash buffer.