Please login to your online account to display your discounted pricing

Superior™ Glove Works Superior Touch™ PU Palm-Coated Dyneema™ Cut-Resistant Gloves

$13.13 - $13.13

Specifications

Color White
Cut Resistance Rating Level 2
Material Dyneema™
For Use With (Application) sharp Steel, Glass, circuit boards, food applications
Glove Cuff Style Knit
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

17-489-615

 
superior glove works ltd
S13SXPUQ7
7 Pair for $13.13
Case of 72 Pair for $878.40

17-489-614

 
superior glove works ltd
S13SXPUQ8
8 Pair for $13.13
Case of 72 Pair for $878.40

17-489-617

 
superior glove works ltd
S13SXPUQ10
10 Pair for $13.13
Case of 72 Pair for $878.40

17-489-618

 
superior glove works ltd
S13SXPUQ9
9 Pair for $13.13
Case of 72 Pair for $878.40
Description & Specifications

Specifications

Color White
Cut Resistance Rating Level 2
Material Dyneema™
For Use With (Application) sharp Steel, Glass, circuit boards, food applications
Glove Cuff Style Knit
Weight Light weight

Polyurethanecoated palm offer protection yet are highly destrous and flexible

  • 13-guage Dyneema is lint-free and provides excellent cut and abrasion resistance
  • Polyurethane provides great grip in wet, oily conditions
  • Snug fitting, breathable and flexible
  • Ergonomically shaped
  • Washable and bleachable (under 145°C)
  • Economy version. 44 grams/pr
  • CFIA approved for food handling