Superior Glove™ Tenactiv™ String Knit Cut-Resistant Gloves
Knit with high-tenacity composite yarns designed to meet a range of applications
$8.31 - $13.66
Specifications
Material Composite Fiber
For Use With (Application) handling sharp objects, Glass plants, construction, automotive industry, aerospace industry, Metal fabrication, Metal stamping
View More Specs
Products 20
Catalog Number Mfr. No. Color Size Price Quantity  
Catalog Number Mfr. No. Color Size Price Quantity  
17-401-240
View Documents
Superior Glove™
S13TAFNT6
Black 6
Pair for $9.11
Case of 240 Pair for $1,951.20
17-401-242
View Documents
Superior Glove™
S13TAFNT8
Black 8
Pair for $9.42
Case of 240 Pair for $2,016.00
17-401-241
View Documents
Superior Glove™
S13TAFNT7
Black 7
Pair for $9.11
Case of 240 Pair for $1,951.20
17-401-243
View Documents
Superior Glove™
S13TAFNT9
Black 9
Pair of 12 for $8.31
Case of 240 Pair for $1,778.40
17-401-244
View Documents
Superior Glove™
S13TAFNT10
Black 10
Pair of 12 for $9.42
Case of 240 Pair for $2,016.00
17-401-245
View Documents
Superior Glove™
S13TAFNT11
Black 11
Pair of 12 for $9.11
Case of 240 Pair for $1,951.20
29-636-3136
View Documents
Superior Glove™
S13TAGPU7
Gray 7
Pair of 12 for $10.55
Case of 240 Pair for $2,258.40
29-636-3135
View Documents
Superior Glove™
S13TAGPU6
Gray 6
Pair of 12 for $10.40
Case of 240 Pair for $2,227.20
29-636-3140
View Documents
Superior Glove™
S13TAGPU11
Gray 11
Pair of 12 for $10.27
Case of 240 Pair for $2,198.40
29-636-3137
View Documents
Superior Glove™
S13TAGPU8
Gray 8
Pair of 12 for $10.27
Case of 240 Pair for $2,198.40
29-636-3139
View Documents
Superior Glove™
S13TAGPU9
Gray 9
Pair of 12 for $10.21
Case of 240 Pair for $2,184.00
17-401-256
View Documents
Superior Glove™
STAFGFNT7
Black 7
Pair of 12 for $13.24
Case of 240 Pair for $2,832.00
29-636-3138
View Documents
Superior Glove™
S13TAGPU10
Gray 10
Pair of 12 for $10.10
Case of 240 Pair for $2,162.40
17-401-255
View Documents
Superior Glove™
STAFGFNT6
Black 6
Pair of 12 for $13.21
Case of 240 Pair for $2,827.20
17-401-254
View Documents
Superior Glove™
S13TAGPU12
Gray 12
Pair of 12 Each for $9.42
Case of 240 Pair for $2,016.00
17-401-257
View Documents
Superior Glove™
STAFGFNT8
Black 8
Pair of 12 Each for $12.02
Case of 240 Pair for $2,572.80
17-401-258
View Documents
Superior Glove™
STAFGFNT9
Black 9
Pair of 12 for $13.24
Case of 240 Pair for $2,832.00
17-401-260
View Documents
Superior Glove™
STAFGFNT11
Black 11
Pair of 12 for $13.24
Case of 240 Pair for $2,832.00
17-401-259
View Documents
Superior Glove™
STAFGFNT10
Black 10
Pair of 12 for $13.66
Case of 240 Pair for $2,923.20
17-401-261
View Documents
Superior Glove™
STAFGFNT12
Black 12
Pair of 12 Each for $13.24
Case of 240 Pair for $2,832.00
Description
Description
  • Continuous filament yarn is stronger than steel on an equal-weight basis
  • 1250 grams of cut protection
  • Lint-freel
Nitrile Palm Coat Style
  • Withstands oils and solvents
  • Touch screen compatible
  • Black
  • Ideal for handling sharp, oily glass or metal
  • ASTM ANSI™ Cut Level 3
Polyurethane-Coated Style
  • Non-shedding
  • High coefficient of friction
  • Color: grey
  • Ideal for those working with circuit boards or any application concerned with contamination
  • ASTM ANSI Cut Level 2
Specifications
Specifications
Composite Fiber
handling sharp objects, Glass plants, construction, automotive industry, aerospace industry, Metal fabrication, Metal stamping
SDS