Superior Glove™ TenActiv™ Composite Knit Cut-Resistant Gloves, Polyurethane Coating
Knit with high-tenacity composite yarns designed to meet a range of applications. Polyurethane coating offers a high coefficient of friction that is non-shedding.
Supplier: Superior Glove™ S13TAGPU11
Description
- Continuous filament yarn is stronger than steel on an equal-weight basis
- 1250 grams of cut protection
- Lint-free
Withstands oils and solvents
Specifications
Polyurethane | |
ANSI A2 | |
Cut-resistant Gloves | |
Handling sharp objects, Glass plants, construction, automotive industry, aerospace industry, Metal fabrication, Metal stamping |
Gray | |
Composite Fiber | |
11 | |
Knit |