Superior Glove™ Tenactiv™ String Knit Cut-Resistant Gloves
Knit with high-tenacity composite yarns designed to meet a range of applications
Manufacturer:  Superior Glove™ S13TAGPU10
 View more versions of this product
Catalog No. 29-636-3138
Description
Description
  • Continuous filament yarn is stronger than steel on an equal-weight basis
  • 1250 grams of cut protection
  • Lint-freel
Nitrile Palm Coat Style
  • Withstands oils and solvents
  • Touch screen compatible
  • Black
  • Ideal for handling sharp, oily glass or metal
  • ASTM ANSI™ Cut Level 3
Polyurethane-Coated Style
  • Non-shedding
  • High coefficient of friction
  • Color: grey
  • Ideal for those working with circuit boards or any application concerned with contamination
  • ASTM ANSI Cut Level 2
Specifications
Specifications
High Tenacity Composite Yarn Fiber
handling sharp objects, Glass plants, construction, automotive industry, aerospace industry, Metal fabrication, Metal stamping
10
Knit
Composite Fiber
Gray