Superior Glove™ Nitrex Cotton Gloves with 3/4 Nitrile Palm Coat
Cut and abarasion resistant with better flexibility
$2.21 - $2.22
Specifications
Coating Material Nitrile
Description Nitrex Cotton
For Use With (Application) Castings, Sheet Metal, Metal Stampings, Brick, Plywood
Puncture Rating ASTM ANSI Puncture 2
Material Cotton
View More Specs
Products 2
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
29-635-8699 View Documents Superior Glove Works Ltd
GNT9
Pair for $2.21
Dozen of 12 Pair for $25.68
29-635-8700 View Documents Superior Glove Works Ltd
GNT10
Pair for $2.22
Dozen of 12 Pair for $25.92
Description
Description
  • Cotton cut-and-sewn support with snug knit wrists
  • Nitrile rubber covers entire palm area and 3/4th of backs
  • Oil and grease resistant
  • ASTM™ puncture level 2 offers low protection against punctures
Castings, Sheet metal, Metal stampings, Brick, Plywood
Compliance
ATSM Puncture Level 2
Specifications
Specifications
Nitrile
Castings, Sheet Metal, Metal Stampings, Brick, Plywood
Cotton
Nitrex Cotton
ASTM ANSI Puncture 2