missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Superior Glove™ Emerald CX™ Nylon/Stainless Steel Knit Cut-Resistant Gloves
Nylon and fine-gauge steel cut-resistant gloves with polyurethane palm coating provides a secure grip, dexterity and touch sensitivity.
Supplier: Superior Glove™ S13KBPU12
Description
- Combination of nylon and fine-gauge steel
- Lint-free nylon is lightweight and tough for better dexterity
- Comfortable with touch sensitivity
- Polyurethane palm coating provides secure grip and tactile feel
- Ideal for glass handling, non-marring gloves will not leave residue, smudges or fingerprints
- Extra-long knit wrists provide extra protection
- Touchscreen compatible
Specifications
ANSI 4 | |
Gray | |
Nylon, Stainless Steel Fiber | |
12 | |
Knit | |
ANSI/ISEA 105-16 Level 2 | |
13 |
Polyurethane | |
ANSI A2 | |
Cut-resistant Gloves | |
HVAC, automotive, sharp Steel, Glass, Metal fabrication, light Metal stamping | |
S13KBPU/12 | |
Emerald CX™ |