missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Super Power Holding Magnets
Available in blocks or buttons.
Manufacturer: Fisher Scientific 711D
Description
- Blocks are 2L × 1W × 0.5 in.H (5 × 2.5 × 1.25cm)
- Buttons are 1.13 in. dia. (2.8cm)
- Box of 40
Specifications
Block magnet display | |
0.5 in. | |
2 in. | |
Plastic | |
2.5cm |
2 x 1 x 0.5 in. (5 x 2.5 x 1.25cm) | |
1.25cm | |
5cm | |
1 in. | |
For science projects, crafts, hobbies and more |